Thermo Scientific Chemicals

Guaiazulene, 99%, Thermo Scientific Chemicals

Catalog number: 120200250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Guaiazulene, 99%, Thermo Scientific Chemicals

Catalog number: 120200250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120200250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
489-84-9
IUPAC Name
1,4-dimethyl-7-(propan-2-yl)azulene
Molecular Formula
C15H18
InChI Key
FWKQNCXZGNBPFD-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C)C2=CC=C(C)C2=C1
Appearance (Color)
Dark blue
Infrared spectrum
Conforms
Appearance (Form)
Low melting crystalline solid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text