Thermo Scientific Chemicals

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals

Catalog number: 120230100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific Chemicals

Catalog number: 120230100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 120230100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50-01-1
IUPAC Name
hydrogen guanidine chloride
Molecular Formula
CH6ClN3
InChI Key
PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(N)=N
Appearance (Form)
Crystalline powder
Water
=<0.3 %
Anticaking agent
=<0.4 % (SiO2)
Appearance (Color)
White to light yellow
Melting point
179°C to 186°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text