DynaMag™-96 Side Skirted Magnet
DynaMag™-96 Side Skirted Magnet
Invitrogen™

DynaMag™-96 Side Skirted Magnet

The DynaMag™-96 Side Skirted magnet is optimized to pull all types of Dynabeads™ (range 1–4.5 μm in diameter) to theRead more
Promo ImagePromotion
Promo code:RDNAP25
Get up to 19% Off Premium DNA Purification Solutions!
Save on our wide range of DNA purification solutions, including column-based extraction kits, magnetic beads, organic extraction reagents, and other essentials.Learn More
Have Questions?
Catalog NumberQuantity
12027Promo Image1 Each
Catalog number 12027
Price (USD)
1,174.65
Online Exclusive
1,236.00
Save 61.35 (5%)
Each
In stock
Add to cart
Quantity:
1 Each
Price (USD)
1,174.65
Online Exclusive
1,236.00
Save 61.35 (5%)
Each
Add to cart
The DynaMag™-96 Side Skirted magnet is optimized to pull all types of Dynabeads™ (range 1–4.5 μm in diameter) to the sides in multi-well plates. The magnet features 6 bar magnets and is compatible with most 96-well skirted PCR-plates, culture plates, 96-well round bottom plates, 96-well flat bottom plates, 24-well plates, 12-well plates, and 6-well plates (not recommended for use with 48-well plates). Working volume per well (5–200 μL). Note that plates may vary, and you should test your plate for compatibility.

Efficient mixing without sample handling
Mixing can be performed by shifting the 96-well plate back and forth from the right-most position to the left-most position.This shifting draws the beads back and forth through the sample and allows mixing to be achieved without handling, promoting faster processing and less sample handing-induced variation.

Handling volume
When used in combination with the DynaMag™-96 Bottom magnet on an automation platform, the two magnets allow for significant volume changes through a workflow.
• High wash volumes: DynaMag™-96 Side magnet and DynaMag™-96 Side Skirted magnet
• Low elution volumes: DynaMag™-96 Bottom magnet

Learn more about Dynabeads™ products
• Find Dynabeads™ products for a whole range of applications.
• Find other magnets for Dynabeads™ separations.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionMagnet
For Use With (Equipment)DynaMag™
Quantity1 Each
Shipping ConditionRoom Temperature
Product LineDYNAL™, Dynabeads™, MPC™
Unit SizeEach
Contents & Storage
Contains one magnet. Store at room temperature.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link