1203-0050PK also known as 1203-0050
Full specifications
Pack of 12 50 mL - Color Clear Description Beaker Griffin Low-Form PMP 50 mL Capacity (Metric) 50 mL Material PMP Graduation Range 10 to 50 mL Increments 5 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 10mL to 50mL in 5mL increments Style Griffin, Low Form Unit Size Pack of 12
1203-0030PK also known as 1203-0030
Full specifications
Pack of 12 30 mL Contact Us
- - Description Beaker Griffin Low-Form PMP 30 mL Capacity (Metric) 30 mL Material PMP Graduation Range 5 to 30 mL Increments 5 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 5mL to 30mL in 5 mL increments Style Griffin, Low Form Unit Size Pack of 12
1203-0050 Full specifications
Case of 36 50 mL - Color Clear Description Beaker Griffin Low-Form PMP 50 mL Capacity (Metric) 50 mL Material PMP Graduation Range 10 to 50 mL Increments 5 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 10mL to 50mL in 5mL increments Style Griffin, Low Form Unit Size Case of 36
1203-0100 Full specifications
Case of 36 100 mL - Color Clear Description Beaker Griffin Low-Form PMP 100 mL Capacity (Metric) 100 mL Material PMP Graduation Range 10 to 100 mL Increments 10 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 10mL to 100mL in 10mL increments Style Griffin, Low Form Unit Size Case of 36
1203-0100PK also known as 1203-0100
Full specifications
Pack of 12 100 mL - Color Clear Description Beaker Griffin Low-Form PMP 100 mL Capacity (Metric) 100 mL Material PMP Graduation Range 10 to 100 mL Increments 10 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 10mL to 100mL in 10mL increments Style Griffin, Low Form Unit Size Pack of 12
1203-0150 Full specifications
Case of 36 150 mL - Capacity (English) 5.072 oz. Color Clear Description Beaker Griffin Low-Form PMP 150 mL Capacity (Metric) 150 mL Material PMP Graduation Range 20 to 150 mL Increments 10 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 20mL to 150mL in 10mL increments Style Griffin, Low Form Unit Size Case of 36
1203-0150PK also known as 1203-0150
Full specifications
Pack of 12 150 mL - Capacity (English) 5.072 oz. Color Clear Description Beaker Griffin Low-Form PMP 150 mL Capacity (Metric) 150 mL Material PMP Graduation Range 20 to 150 mL Increments 10 mL Quantity 12/Pk.; 36/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 20mL to 150mL in 10mL increments Style Griffin, Low Form Unit Size Pack of 12
1203-0250 Full specifications
Case of 24 250 mL - Color Clear Description Beaker Griffin Low-Form PMP 250 mL Capacity (Metric) 250 mL Material PMP Graduation Range 25 to 250 mL Increments 25 mL Quantity 6/Pk.; 24/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 25mL to 250mL in 25mL increments Style Griffin, Low Form Unit Size Case of 24
1203-0250PK also known as 1203-0250
Full specifications
Pack of 6 250 mL - Color Clear Description Beaker Griffin Low-Form PMP 250 mL Capacity (Metric) 250 mL Material PMP Graduation Range 25 to 250 mL Increments 25 mL Quantity 6/Pk.; 24/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 25mL to 250mL in 25mL increments Style Griffin, Low Form Unit Size Pack of 6
1203-0400 Full specifications
Case of 24 400 mL - Color Clear Description Beaker Griffin Low-Form PMP 400 mL Capacity (Metric) 400 mL Material PMP Graduation Range 50 to 400 mL Increments 25 mL Quantity 6/Pk.; 24/Cs. Type Beaker Autoclavable Autoclavable Disposable Reusable Graduated Yes Certifications/Compliance ISO/DIN 7056 Scale 50mL to 400mL in 25mL increments Style Griffin, Low Form Unit Size Case of 24