1-Heptanol, 98%
1-Heptanol, 98%
1-Heptanol, 98%
Thermo Scientific Chemicals

1-Heptanol, 98%

CAS: 111-70-6 | C7H16O | 116.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 120362500
Price (EUR)
24,31
Online Exclusive
28,60
Save 4,29 (15%)
Each
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
24,31
Online Exclusive
28,60
Save 4,29 (15%)
Each
Add to cart
Chemical Identifiers
CAS111-70-6
IUPAC Nameheptan-1-ol
Molecular FormulaC7H16O
InChI KeyBBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILESCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Bromine number=<0.1 g Br/100 g
Infrared spectrumConforms
Acid number=<0.5 mg KOH/g
Residue after evaporation=<0.01 %
Color scale=<30 APHA
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467533Certificate of AnalysisNov 11, 2024120360025, 120362500, 120360010
A0466582Certificate of AnalysisOct 03, 2024120360025, 120362500, 120360010
A0457386Certificate of AnalysisNov 08, 2023120360025, 120362500, 120360010
A0451613Certificate of AnalysisMay 02, 2023120360025, 120362500, 120360010
A0397348Certificate of AnalysisDec 15, 2022120360025, 120362500, 120360010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H319 - Causes serious eye irritation

EU Specific Hazard Statements

Combustible liquid

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

Share catalog number, name or link

1x1 image pixel for data collection