Thermo Scientific Chemicals

2-Heptanol, 99+%, Thermo Scientific Chemicals

Catalog number: 120370010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

2-Heptanol, 99+%, Thermo Scientific Chemicals

Catalog number: 120370010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 120370010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
543-49-7
IUPAC Name
heptan-2-ol
Molecular Formula
C7H16O
InChI Key
CETWDUZRCINIHU-UHFFFAOYNA-N
SMILES
CCCCCC(C)O
Appearance (Color)
Clear colorless
GC
>=99.0 %
Specific gravity
(20°C) 0.816 to 0.819
Appearance (Form)
Liquid
Refractive index
1.4200 to 1.4220 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text