1-Heptene, 98%, Thermo Scientific Chemicals
1-Heptene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Heptene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 120390250
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
Add to cart
1-Heptene, 98%, Thermo Scientific Chemicals
Catalog number120390250
Price (USD)/ Each
100.65
Online exclusive
112.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namehept-1-ene
Molecular FormulaC7H14
InChI KeyZGEGCLOFRBLKSE-UHFFFAOYSA-N
SMILESCCCCCC=C
Molecular Weight (g/mol)98.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only