Gibco™

Hybridoma-SFM

Catalog number: 12045076
Have Questions?
Gibco™

Hybridoma-SFM

Catalog number: 12045076
Have Questions?
Catalog Number
12045076
Unit Size
1,000 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
120450761,000 mL
-
12045084500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Hybridoma-SFM is a serum-free, very low-protein medium specifically developed to support the growth of hybridomas for antibody production. Hybridoma-SFM features:

• Ability to support suspension and stationary hybridomas and myelomas
• Complete, ready-to-use formulation
• Serum-free, very low-protein formulation

Ability to support suspension and stationary hybridomas and myelomas
Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Hybridoma-SFM provides high cell densities and IgG production. Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate.

Complete, ready-to-use formulation
Hybridoma-SFM is a complete medium that is ready to use. No supplementation is required.

Serum-free, very low-protein formulation
Hybridoma-SFM is a serum-free, very low-protein formulation, allowing for easier purification of your protein of interest. Hybridoma-SFM contains <20 μg/mL total protein.

Product use
Customers using Gibco™ Hybridoma-SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
Hybridoma-SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Hybridomas
Classification
Low Protein, Serum-free
Form
Liquid
Product Type
Serum Free Medium (SFM)
Serum Level
Serum-free
With Additives
Glutamine, Phenol Red
Green Features
Sustainable packaging
Product Line
Gibco™
Quantity
1000 mL
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text