Thermo Scientific Chemicals

n-Hexadecane, 99%, pure, Thermo Scientific Chemicals

Catalog number: 120460025
2.5 L, Each
Thermo Scientific Chemicals

n-Hexadecane, 99%, pure, Thermo Scientific Chemicals

Catalog number: 120460025
2.5 L, Each
Quantity
Catalog number: 120460025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
544-76-3
IUPAC Name
hexadecane
Molecular Formula
C16H34
InChI Key
DCAYPVUWAIABOU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCC
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Liquid or solid
Refractive index
1.4330 to 1.4350 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text