n-Hexadecane, 99%, pure, Thermo Scientific Chemicals
n-Hexadecane, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Hexadecane, 99%, pure, Thermo Scientific Chemicals

n-Hexadecane, 99%, C16H34, CAS Number-544-76-3, n-cetane, n-hexadecane, hexadecan, hexadekan, cetan, cetane, ccris 5833, zetan, pentadecane, methyl, unii-f8z00shp6q, 2.5L, 98.5% min. (GC), 01,172, 287.0 deg.C, 100.0, CHEBI:45296, Colorless, 0.7730g/mL, 208-878-9, 135 deg.C, 226.44, Pure
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
10 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 120461000
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
Add to cart
n-Hexadecane, 99%, pure, Thermo Scientific Chemicals
Catalog number120461000
Price (USD)/ Each
80.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
94.20 
Save 13.55 (14%)
-
Add to cart
Chemical Identifiers
CAS544-76-3
IUPAC Namehexadecane
Molecular FormulaC16H34
InChI KeyDCAYPVUWAIABOU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid or solid
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only