1-Hexadecylamine, 90%, Thermo Scientific Chemicals
1-Hexadecylamine, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hexadecylamine, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Plastic drum
Catalog number 120510025
Price (USD)/ Each
203.00
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
203.00
Add to cart
1-Hexadecylamine, 90%, Thermo Scientific Chemicals
Catalog number120510025
Price (USD)/ Each
203.00
-
Add to cart
Chemical Identifiers
CAS143-27-1
IUPAC Namehexadecan-1-amine
Molecular FormulaC16H35N
InChI KeyFJLUATLTXUNBOT-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point38°C to 47°C
Appearance (Form)Low melting crystalline mass and/or chunks
GC>=88 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only