Thermo Scientific Chemicals

1-Hexadecylamine, 90%, Thermo Scientific Chemicals

Catalog number: 120515000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Hexadecylamine, 90%, Thermo Scientific Chemicals

Catalog number: 120515000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120515000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-27-1
IUPAC Name
hexadecan-1-amine
Molecular Formula
C16H35N
InChI Key
FJLUATLTXUNBOT-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCN
Appearance (Color)
White
Melting point
38°C to 47°C
Appearance (Form)
Low melting crystalline mass and/or chunks
GC
>=88 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text