Thermo Scientific Chemicals

2,4-Hexadienoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 120535000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

2,4-Hexadienoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 120535000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 120535000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-44-1
IUPAC Name
(2E,4E)-hexa-2,4-dienoic acid
Molecular Formula
C6H8O2
InChI Key
WSWCOQWTEOXDQX-MQQKCMAXSA-N
SMILES
C\C=C\C=C\C(O)=O
Appearance (Color)
White to cream to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
132°C to 135°C
Titration with NaOH
98.5 to 101.5 % (on anhydrous substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text