Thermo Scientific Chemicals

1,1,1,3,3,3-Hexamethyldisilazane, 98%, Thermo Scientific Chemicals

Catalog number: 120580100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexamethyldisilazane, 98%, Thermo Scientific Chemicals

Catalog number: 120580100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 120580100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
999-97-3
IUPAC Name
bis(trimethylsilyl)amine
Molecular Formula
C6H19NSi2
InChI Key
FFUAGWLWBBFQJT-UHFFFAOYSA-N
SMILES
C[Si](C)(C)N[Si](C)(C)C
Color scale
=<30 APHA
GC
>=97.5 %
Refractive index
1.4060 to 1.4090 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text