Macrophage-SFM is a serum-free medium designed for the culture of human peripheral blood monocytes and macrophages (1).Contains L-glutamine, but noRead more
Have Questions?
Catalog Number
Quantity
12065074
500 mL
Catalog number 12065074
Price (USD)
165.65
Online Exclusive
180.00
Save 14.35 (8%)
Each
Add to cart
Quantity:
500 mL
Price (USD)
165.65
Online Exclusive
180.00
Save 14.35 (8%)
Each
Add to cart
Macrophage-SFM is a serum-free medium designed for the culture of human peripheral blood monocytes and macrophages (1).
Contains L-glutamine, but no antibiotics.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMacrophages, Monocytes
Concentration1 X
Endotoxin Level≤1 EU/mL, ≤ 1 EU/mL
Included AntibioticsNo antibiotics
Product TypeSerum Free Medium (SFM)
Quantity500 mL
Shipping ConditionRoom Temperature
ClassificationSerum-free
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelSerum-free
SterilitySterile-filtered
With AdditivesGlutamine
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.