Thermo Scientific Chemicals

Hexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 120700025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Hexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 120700025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 120700025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
142-62-1
IUPAC Name
hexanoic acid
Molecular Formula
C6H12O2
InChI Key
FUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILES
CCCCCC(O)=O
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Appearance (Form)
Liquid
Refractive index
1.4150 to 1.4180 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text