Hexanoic acid, 99%
Hexanoic acid, 99%
Hexanoic acid, 99%
Thermo Scientific Chemicals

Hexanoic acid, 99%

CAS: 142-62-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 120705000
Price (USD)
61.65
Special offer
Online exclusive
Ends: 26-Sep-2025
76.10
Save 14.45 (19%)
Each
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
61.65
Special offer
Online exclusive
Ends: 26-Sep-2025
76.10
Save 14.45 (19%)
Each
Add to cart
Chemical Identifiers
CAS142-62-1
IUPAC Namehexanoic acid
Molecular FormulaC6H12O2
InChI KeyFUZZWVXGSFPDMH-UHFFFAOYSA-N
SMILESCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4150 to 1.4180 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link