1-Hexene, 99%, AcroSeal™, Thermo Scientific Chemicals
1-Hexene, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hexene, 99%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 120751000
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
1-Hexene, 99%, AcroSeal™, Thermo Scientific Chemicals
Catalog number120751000
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.3867 to 1.3887 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only