5-Hexen-2-one, 98%, Thermo Scientific Chemicals
5-Hexen-2-one, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hexen-2-one, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
20 mL
100 mL
Packaging:
Glass bottle
Catalog number 120780050
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.10 
Save 4.45 (15%)
-
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.10 
Save 4.45 (15%)
Add to cart
5-Hexen-2-one, 98%, Thermo Scientific Chemicals
Catalog number120780050
Price (USD)/ Each
24.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
29.10 
Save 4.45 (15%)
-
Add to cart
Chemical Identifiers
CAS109-49-9
IUPAC Namehex-5-en-2-one
Molecular FormulaC6H10O
InChI KeyRNDVGJZUHCKENF-UHFFFAOYSA-N
SMILESCC(=O)CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only