Hexyl alcohol, 98%, pure, Thermo Scientific Chemicals
Hexyl alcohol, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexyl alcohol, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Metal Drum
Catalog number 120790250
Price (USD)/ Each
565.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
665.00 
Save 99.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)/ Each
565.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
665.00 
Save 99.35 (15%)
Add to cart
Hexyl alcohol, 98%, pure, Thermo Scientific Chemicals
Catalog number120790250
Price (USD)/ Each
565.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
665.00 
Save 99.35 (15%)
-
Add to cart
Chemical Identifiers
CAS111-27-3
IUPAC Namehexan-1-ol
Molecular FormulaC6H14O
InChI KeyZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILESCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Color scale=<10 APHA
GC>=97.5 %
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
Acid value=<0.02 mg KOH/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only