Thermo Scientific Chemicals

DL-Homocysteinethiolactone hydrochloride, 99%

Catalog number: 120831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Homocysteinethiolactone hydrochloride, 99%

Catalog number: 120831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6038-19-3
IUPAC Name
hydrogen 3-aminothiolan-2-one chloride
Molecular Formula
C4H8ClNOS
InChI Key
ZSEGSUBKDDEALH-UHFFFAOYNA-N
SMILES
[H+].[Cl-].NC1CCSC1=O
Titration with HClO4
>=98.5 %
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text