Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
Packaging:
Plastic bottle
Catalog number 120860010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS68131-04-4
IUPAC Namedisodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Molecular FormulaC9H8Na2O4
InChI KeyHMGUIQPKFUZDPV-UHFFFAOYNA-L
SMILES[Na+].[Na+].[O-]C(=O)C1C2CC(C=C2)C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Assay45 to 70 % (as Humic acid)
Appearance (Color)Black
Infrared spectrumConforms
Loss on drying14 % to 25 % (105°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only