Thermo Scientific Chemicals

Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals

Catalog number: 120861000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Humic acid, sodium salt, 45-70%, Thermo Scientific Chemicals

Catalog number: 120861000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 120861000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68131-04-4
IUPAC Name
disodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Molecular Formula
C9H8Na2O4
InChI Key
HMGUIQPKFUZDPV-UHFFFAOYNA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1C2CC(C=C2)C1C([O-])=O
Appearance (Form)
Crystalline powder
Assay
45 to 70 % (as Humic acid)
Appearance (Color)
Black
Infrared spectrum
Conforms
Loss on drying
14 % to 25 % (105°C, 4 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text