Thermo Scientific Chemicals

2-Hydrazinopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 120870010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Hydrazinopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 120870010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 120870010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4930-98-7
IUPAC Name
2-hydrazinylpyridine
Molecular Formula
C5H7N3
InChI Key
NWELCUKYUCBVKK-UHFFFAOYSA-N
SMILES
NNC1=CC=CC=N1
Appearance (Form)
Crystals or crystalline mass
Melting point
43°C to 47°C
Infrared spectrum
Conforms
Appearance (Color)
White to orange-brown
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text