Hydroquinone, 99%, Thermo Scientific Chemicals
Hydroquinone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroquinone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
10 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 120910020
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
Add to cart
Hydroquinone, 99%, Thermo Scientific Chemicals
Catalog number120910020
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namebenzene-1,4-diol
Molecular FormulaC6H6O2
InChI KeyQIGBRXMKCJKVMJ-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C=C1
Molecular Weight (g/mol)110.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point170°C to 175°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals or needles
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only