2'-Hydroxyacetophenone, 99%, Thermo Scientific Chemicals
2'-Hydroxyacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Hydroxyacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 120931000
Price (USD)/ Each
28.65
Online exclusive
31.40 
Save 2.75 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.40 
Save 2.75 (9%)
Add to cart
2'-Hydroxyacetophenone, 99%, Thermo Scientific Chemicals
Catalog number120931000
Price (USD)/ Each
28.65
Online exclusive
31.40 
Save 2.75 (9%)
-
Add to cart
Chemical Identifiers
CAS118-93-4
IUPAC Name1-(2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O2
InChI KeyJECYUBVRTQDVAT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to brown
Infrared spectrumConforms
Refractive index1.5580 to 1.5600 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only