3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
Thermo Scientific Chemicals

3-Hydroxybenzaldehyde, 98.5%

CAS: 100-83-4 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 120961000
Price (USD)
57.65
Special offer
Online exclusive
67.90
Save 10.25 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
67.90
Save 10.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS100-83-4
IUPAC Name3-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyIAVREABSGIHHMO-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point101°C to 105°C
Appearance (Color)Light yellow to brown
Infrared spectrumConforms
GC>=98.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470036Certificate of AnalysisMay 17, 2025120965000, 120961000
A0408463Certificate of AnalysisDec 13, 2022120965000, 120961000
A0436052Certificate of AnalysisJan 11, 2022120965000, 120961000
A0425579Certificate of AnalysisJan 06, 2021120965000, 120961000
A0422012Certificate of AnalysisAug 26, 2020120965000, 120961000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection