Thermo Scientific Chemicals

4-Hydroxybenzoic Acid, 99+%, Thermo Scientific Chemicals

Catalog number: 120990010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Hydroxybenzoic Acid, 99+%, Thermo Scientific Chemicals

Catalog number: 120990010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 120990010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-96-7
IUPAC Name
4-hydroxybenzoic acid
Molecular Formula
C7H6O3
InChI Key
FJKROLUGYXJWQN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(O)C=C1
Appearance (Form)
Crystalline powder
HPLC
>=99.0 %
Infrared spectrum
Conforms
Titration with NaOH
>=99.0 %
Appearance (Color)
White to cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text