Thermo Scientific Chemicals

3-Hydroxybenzyl Alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 121025000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxybenzyl Alcohol, 97%, Thermo Scientific Chemicals

Catalog number: 121025000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121025000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-24-6
IUPAC Name
3-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
OKVJCVWFVRATSG-UHFFFAOYSA-N
SMILES
OCC1=CC=CC(O)=C1
Appearance (Form)
Crystalline powder or granular powder
Melting point
64°C to 71°C
Appearance (Color)
White to beige to brown
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text