Thermo Scientific Chemicals

3-Hydroxybutyric acid, 95%, Thermo Scientific Chemicals

Catalog number: 121050100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxybutyric acid, 95%, Thermo Scientific Chemicals

Catalog number: 121050100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121050100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
300-85-6
IUPAC Name
3-hydroxybutanoic acid
Molecular Formula
C4H8O3
InChI Key
WHBMMWSBFZVSSR-UHFFFAOYNA-N
SMILES
CC(O)CC(O)=O
Appearance (Form)
Solid or liquid
Titration with NaOH
94 to 106 %
Appearance (Color)
Colorless to white to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text