Thermo Scientific Chemicals

m-Hydroxycinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 121080250
25 g, Each
Thermo Scientific Chemicals

m-Hydroxycinnamic acid, 99%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 121080250
25 g, Each
Quantity
Catalog number: 121080250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14755-02-3
IUPAC Name
(2E)-3-(3-hydroxyphenyl)prop-2-enoate
Molecular Formula
C9H7O3
InChI Key
KKSDGJDHHZEWEP-SNAWJCMRSA-M
SMILES
OC1=CC=CC(\C=C\C([O-])=O)=C1
Appearance (Form)
Crystalline powder
HPLC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
Beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text