p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 121090250
Price (USD)/ Each
91.65
Online exclusive
108.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
91.65
Online exclusive
108.00 
Save 16.35 (15%)
Add to cart
p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Catalog number121090250
Price (USD)/ Each
91.65
Online exclusive
108.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O3
InChI KeyNGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILESOC(=O)C=CC1=CC=C(O)C=C1
Molecular Weight (g/mol)164.16
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to beige to green to light brown
HPLC>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only