Thermo Scientific Chemicals

p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 121091000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

p-Hydroxycinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 121091000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 121091000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
501-98-4
IUPAC Name
(2E)-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular Formula
C9H8O3
InChI Key
NGSWKAQJJWESNS-ZZXKWVIFSA-N
SMILES
OC(=O)C=CC1=CC=C(O)C=C1
Infrared spectrum
Conforms
Appearance (Color)
White to yellow to beige to green to light brown
HPLC
>=97.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text