Thermo Scientific Chemicals

7-Hydroxycoumarin, 99%, Thermo Scientific Chemicals

Catalog number: 121111000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

7-Hydroxycoumarin, 99%, Thermo Scientific Chemicals

Catalog number: 121111000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 121111000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-35-6
IUPAC Name
7-hydroxy-2H-chromen-2-one
Molecular Formula
C9H6O3
InChI Key
ORHBXUUXSCNDEV-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC(=O)OC2=C1
Infrared spectrum
Conforms
HPLC
>=98.5 %
Appearance (Form)
Powder or crystals and/or agglomerates
Melting point
228°C to 234°C
Appearance (Color)
White to yellow to beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text