Thermo Scientific Chemicals

Diethylene glycol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 121160025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Diethylene glycol, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 121160025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 121160025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-46-6
IUPAC Name
2-(2-hydroxyethoxy)ethan-1-ol
Molecular Formula
C4H10O3
InChI Key
MTHSVFCYNBDYFN-UHFFFAOYSA-N
SMILES
OCCOCCO
Appearance (Form)
Clear viscous liquid
Water
=<0.2 %
Ash
=<0.01 %
Infrared spectrum
Conforms
Refractive index
1.4450 to 1.4480 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text