9-Hydroxyfluorene, 97%
9-Hydroxyfluorene, 97%
9-Hydroxyfluorene, 97%
9-Hydroxyfluorene, 97%
Thermo Scientific Chemicals

9-Hydroxyfluorene, 97%

CAS: 1689-64-1 | C13H10O | 182.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12123025025 gGlass bottle
Catalog number 121230250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1689-64-1
IUPAC Name9H-fluoren-9-ol
Molecular FormulaC13H10O
InChI KeyAFMVESZOYKHDBJ-UHFFFAOYSA-N
SMILESOC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting point152°C to 158°C
Appearance (Form)Crystalline powder
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link