5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals
5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 121282500
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
Add to cart
5-Hydroxyindole-3-acetic Acid, 99%, Thermo Scientific Chemicals
Catalog number121282500
Price (USD)/ Each
43.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
50.80 
Save 7.15 (14%)
-
Add to cart
Chemical Identifiers
CAS54-16-0
IUPAC Name2-(5-hydroxy-1H-indol-3-yl)acetate
Molecular FormulaC10H8NO3
InChI KeyDUUGKQCEGZLZNO-UHFFFAOYSA-M
SMILESOC1=CC=C2NC=C(CC([O-])=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Color)Beige
Melting point162°C to 165°C
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only