Thermo Scientific Chemicals

4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 121305000
500g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 121305000
500g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 121305000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
56-04-2
IUPAC Name
6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Molecular Formula
C5H6N2OS
InChI Key
HWGBHCRJGXAGEU-UHFFFAOYSA-N
SMILES
CC1=CC(=O)NC(=S)N1
Appearance (Color)
White to light yellow to beige
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text