Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 121351000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 121351000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121351000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
121-34-6
IUPAC Name
4-hydroxy-3-methoxybenzoic acid
Molecular Formula
C8H8O4
InChI Key
WKOLLVMJNQIZCI-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1O)C(O)=O
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.0 %
Melting point
208°C to 214°C
Appearance (Color)
White to yellow to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text