4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 121370010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7149-10-2
IUPAC Name4-(aminomethyl)-2-methoxyphenol hydrochloridyl
Molecular FormulaC8H11ClNO2
InChI KeyLOYPVODLNGWOLM-UHFFFAOYSA-N
SMILES[Cl].COC1=CC(CN)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only