5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
Thermo Scientific Chemicals

5-(Hydroxymethyl)furfural, 98%

CAS: 67-47-0 | C6H6O3 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 121460010
Price (USD)
60.50
Each
In stock
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.50
Each
Add to cart
Chemical Identifiers
CAS67-47-0
IUPAC Name5-(hydroxymethyl)furan-2-carbaldehyde
Molecular FormulaC6H6O3
InChI KeyNOEGNKMFWQHSLB-UHFFFAOYSA-N
SMILESOCC1=CC=C(O1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow to orange to brown
Appearance (Form)Liquid or crystalline powder and/or chunks or
Appearance (Form)fused solid
Melting point28°C to 34°C
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection