Thermo Scientific Chemicals

5-(Hydroxymethyl)furfural, 98%, Thermo Scientific Chemicals

Catalog number: 121460250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

5-(Hydroxymethyl)furfural, 98%, Thermo Scientific Chemicals

Catalog number: 121460250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121460250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-47-0
IUPAC Name
5-(hydroxymethyl)furan-2-carbaldehyde
Molecular Formula
C6H6O3
InChI Key
NOEGNKMFWQHSLB-UHFFFAOYSA-N
SMILES
OCC1=CC=C(O1)C=O
Appearance (Color)
Colorless to white to yellow to orange to brown
Appearance (Form)
Liquid or crystalline powder and/or chunks or
Appearance (Form)
fused solid
Infrared spectrum
Conforms
Melting point
28°C to 34°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text