N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals
N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(Hydroxymethyl)phthalimide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 121491000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS118-29-6
IUPAC Name2-(hydroxymethyl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC9H7NO3
InChI KeyMNSGOOCAMMSKGI-UHFFFAOYSA-N
SMILESOCN1C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Sulfated ash=<0.1 %
Melting point140°C to 145°C
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only