Thermo Scientific Chemicals

3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals

Catalog number: 121550050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals

Catalog number: 121550050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121550050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
118-71-8
IUPAC Name
3-hydroxy-2-methyl-4H-pyran-4-one
Molecular Formula
C6H6O3
InChI Key
XPCTZQVDEJYUGT-UHFFFAOYSA-N
SMILES
CC1=C(O)C(=O)C=CO1
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
GC
>=98.5 %
Lead (Pb)
=<10 ppm
Melting point
160°C to 164°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text