3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals
3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxy-2-methyl-4-pyrone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 121555000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS118-71-8
IUPAC Name3-hydroxy-2-methyl-4H-pyran-4-one
Molecular FormulaC6H6O3
InChI KeyXPCTZQVDEJYUGT-UHFFFAOYSA-N
SMILESCC1=C(O)C(=O)C=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=98.5 %
Lead (Pb)=<10 ppm
Melting point160°C to 164°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only