3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxy-4-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 121671000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS619-14-7
IUPAC Name3-hydroxy-4-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyXLDLRRGZWIEEHT-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(C(O)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<0.5 %
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Color)Yellow to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only