Thermo Scientific Chemicals

4-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 121711000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 121711000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121711000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
156-38-7
IUPAC Name
2-(4-hydroxyphenyl)acetic acid
Molecular Formula
C8H8O3
InChI Key
XQXPVVBIMDBYFF-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(O)C=C1
Appearance (Form)
Powder or crystals
Melting point
148°C to 155°C
HPLC
>=97.5 %
Appearance (Color)
White to cream to light yellow to light brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text