Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline, 99+%

Catalog number: 121785000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

trans-4-Hydroxy-L-proline, 99+%

Catalog number: 121785000
500 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 121785000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-35-4
IUPAC Name
4-hydroxypyrrolidine-2-carboxylic acid
Molecular Formula
C5H9NO3
InChI Key
PMMYEEVYMWASQN-UHFFFAOYNA-N
SMILES
OC1CNC(C1)C(O)=O
Appearance (Color)
White
Titration with HClO4
99.0 to 101.0 %
Sulfated ash
=<0.1 %
Ammonium (NH4)
=<200 ppm
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text