PureLink™ RNA Mini Kit
PureLink™ RNA Mini Kit
Invitrogen™
PureLink™ RNA Mini Kit
The PureLink RNA Mini Kit is a column-based kit used to isolate high-quality total RNA from a wide variety of sample types in 20 minutes using standard laboratory equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct Type
1218302010 PrepsRNA Mini Kit
12183018A50 PrepsRNA Mini Kit
12183025250 PrepsRNA Mini Kit
A2983950 CartridgesRNA Mini Columns
1218302650 CartridgesHomogenizer
Catalog number 12183020
Price (USD)
166.65
Online Exclusive
175.00
Save 8.35 (5%)
Each
-
Add to cart
Quantity:
10 Preps
Product Type:
RNA Mini Kit
Price (USD)
166.65
Online Exclusive
175.00
Save 8.35 (5%)
Each
Add to cart
Ask our AI about this Product
The PureLink RNA Mini Kit is a column-based kit used to isolate high-quality total RNA from a wide variety of sample types in 20 minutes using standard laboratory equipment. The kit includes RNase-free lysis and wash solutions that protect RNA from RNases while liberating the RNA from DNA, proteins, and other cellular debris. The fast spin-column workflow is ideal for processing low to mid-throughput batch sizes. The advanced PureLink RNA Mini spin column design allows for maximum sample input (200 mg of tissue) and RNA recovery (up to 1000 μg). This means that users can process both small and large sample sizes with the same RNA isolation kit.
  • Process sample inputs from 5–200 mg of animal/plant tissue or from 0.5 x 106 to 108 tissue culture cells with one spin column in 20 minutes
  • Isolate more RNA than competitor mini spin columns: up to 1000 μg of total RNA from one column
  • Use isolated RNA in most applications without the need for further DNase treatment (very low minimal residual DNA carry over)
  • Flexible–optional on-column or post-isolation DNase treatment is available with PureLink DNase Set (Cat. No. 12185010) for easy removal of residual DNA
Extra-large binding capacity enables rapid RNA purification using standard laboratory equipment
The PureLink RNA Mini Kit provides rapid purification of total RNA from a wide range of cells and tissue types to yield up to 1000 μg of purified RNA from a single extraction (see Figure 1). High-quality total RNA can be obtained from mini- to midi-prep amounts of starting material with just trace amounts of residual genomic DNA contamination. The extra-large binding capacity enables one kit to handle most RNA isolation needs in a quick 20 minute protocol without the need for special sample processing instruments.
Easy, optional, on-column DNase treatment for sensitive applications
The PureLink RNA Mini Kit columns are highly efficient for isolating high quality total RNA while removing the majority of genomic DNA. In general, most applications requiring RNA from animal tissue or mammalian cell lines do not require additional DNase treatment. However, some applications such as gene expression analysis by qRT-PCR without intron-spanning primers or working with samples from organisms with very small or no introns may require more complete removal of residual contaminating DNA. The PureLink DNase Set (12185010) allows for convenient on-column digestion of DNA during the RNA isolation protocol. Treating with DNase while 'on-column' is easier and allows higher RNA recovery than treating with DNase after the RNA has been isolated. The PureLink DNase Set can also be used to remove residual DNA from RNA that has been previously purified. Both the 'on-column' and post-RNA purification workflows are options available with the PureLink DNase Set.
Simplified RNA purification
The PureLink RNA Mini Kit utilizes guanidine-isothiocyanate lysis buffer to protect the RNA during the isolation steps. Special RNase-free reagents combined with certified RNase-free silica membranes in a unique column configuration allow for a safe and easy procedure that can typically be completed in less than 20 minutes without the need of hazardous phenol/chloroform extraction, CsCl centrifugation, or LiCl or alcohol precipitation.
The PureLink RNA Mini Kit is recommended for use with the Homogenizer (Cat. No. 12183026), designed to homogenize cell or tissue lysates via centrifugation, prior to nucleic acid purification. The Homogenizer is especially effective for clarifying particulates from plant tissues.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Elution Volume30–300 μL
Final Product TypeTotal RNA
For Use With (Application)RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product TypeRNA Mini Kit
Purification Time20 min. (After sample homogenization)
Quantity10 Preps
ScaleMini
Shipping ConditionRoom Temperature
Starting Material AmountBacteria: ≤109 cells, Blood: ≤0.2 mL, Cells: ≤5 x 107, Plant: ≤250 mg, Tissue: ≤200 mg, Yeast: ≤5 x 108 cells, Enzymatic reactions: ≤1.2 mL
YieldUp to 350 μg
Isolation TechnologySilica Spin Column
Sample TypeBacteria, Blood, Cells, Plant, Tissue, Yeast, Enzymatic Reactions
Unit SizeEach
Contents & Storage
• 20 mL Lysis Buffer; store at room temperature
• 10 mL Wash Buffer I; room temperature
• 4 mL Wash Buffer II; room temperature
• 3 mL RNase-free Water; room temperature
• 10 PureLink Spin Cartridges with Collection Tubes; room temperature
• 10 Collection Tubes; room temperature
• 10 Recovery Tubes; room temperature
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Figures
Frequently asked questions (FAQs)
The presence of ethanol or salt in the purified RNA can inhibit downstream enzymatic reactions. Ensure that you are using the correct order of wash buffers in the kit for washing, and that Wash Buffer II is discarded in the flow-through. Place the spin cartridge into the wash tube and centrifuge the spin cartridge at maximum speed for 2-3 minutes to completely dry the cartridge.
The RNA could have been contaminated with RNase. Ensure that you are using RNase-free equipment and change gloves frequently. Improper handling can also result in RNA degradation. Ensure samples are processed immediately, and that the lysis is performed quickly after adding the lysis buffer. Lastly, tissues rich in RNase (such as rat pancreas) may require the addition of RNase inhibitors or inactivators to protect the RNA from degradation, or a larger volume of lysis buffer.
Low RNA yield can occur due to the following:
- Incomplete lysis and homogenization: ensure that 10 µL of 2-mercaptoethanol was added per milliliter of lysis buffer, perform all steps at room temperature, decrease the amount of starting material used, use proper homogenization methods, and/or cut tissue samples into smaller pieces to ensure complete tissue immersion in the lysis buffer.
- Poor quality starting material: use fresh samples and process immediately after collection.
- Ethanol may not have been added to Wash Buffer II.
- Incorrect elution conditions may have been used: Add RNase-free water and incubate for 1 minute before centrifugation, following the recommendations for elution in the manual. You can also perform a second elution step to recover more RNA.
Incomplete homogenization or dispersal of precipitate after ethanol addition can lead to clogging of the RNA spin cartridge. Clear the homogenate and remove any particulate or viscous material by centrifugation. Completely disperse any precipitate that forms after adding ethanol to the homogenate. Load only the supernatant onto the RNA spin cartridge to avoid clogging.
The PureLink RNA Mini Kit provides rapid column-based purification of total RNA, without organic lysis (phenol/chloroform). You can obtain up to 1,000 µg of purified RNA from a single extraction. The RiboPure RNA Purification Kits combine a phenol/guanidine thiocyanate solution with a glass-fiber filter purification method.
Share catalog number, name or link
1x1 image pixel for data collection