Thermo Scientific Chemicals

Allopurinol, 98%

Catalog number: 121831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Allopurinol, 98%

Catalog number: 121831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 121831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
315-30-0
IUPAC Name
1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Formula
C5H4N4O
InChI Key
OFCNXPDARWKPPY-UHFFFAOYSA-N
SMILES
O=C1N=CN=C2NNC=C12
Infrared spectrum
Conforms
Appearance (Color)
White to almost white
Loss on drying
=<0.5 % (1 g, 105°C)
Appearance (Form)
Powder
Titration with NaOH
>=97.5 % (on dried substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text